Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FTNVTLSSIK ETRGLHQQDC VHEAEEGSKL KILGPFSNFF ARNLLARKQS ARLDKHNDLG WKLFGKAPLR ENAQKDSKRI QKEYED
Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
Plays a role in the regulation of starvation-induced autophagosome formation (PubMed:22613832). Together with the TRAPPIII complex, regulates a constitutive trafficking step from peripheral recycling endosomes to the early Golgi, maintaining the cycling pool of ATG9 required for initiation of autophagy. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TBC1 domain family member 14; TBC1 domain family, member 14
Gene Aliases: KIAA1322; TBC1D14
UniProt ID: (Human) Q9P2M4
Entrez Gene ID: (Human) 57533
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support