The recommended shelf life for this product is 1 year from date of receipt.
Acts as a regulator of transcriptional activation. Plays a role in the inducible expression of cytokine genes in T-cells, especially in the induction of the IL-2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GAP for RAB7; GTPase-activating protein RAB7; lymphocyte antigen 6, locus D ligand; TBC1 domain family member 15; TBC1 domain family, member 15; Tre-2-budding uninhibited by benzimidazole-cell division cycle 16 domain, domain family member 15; Tre2/Bub2/Cdc16 domain family member 15
Gene Aliases: 4432405K22Rik; AU022358; AU023730; AW557774; Ly6dl; RAB7-GAP; TBC1D15
UniProt ID: (Human) Q8TC07, (Mouse) Q9CXF4
Entrez Gene ID: (Human) 64786, (Mouse) 66687
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support