Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALRSAQGDG PTSGHWDGGA EKADFNAKRK KKVAEIHQAL NSDPTDVAAL RRMAISEGGL LTDEIRRKVW PKLLNVNAND PPPISGKNLR QMSKDYQQVL LDVRRSLRRF PPGMPEEQRE GLQEELIDII LLILERNPQL HYYQGYHDIV VTFLLVVGER LATSLVEKLS THHLRDFMDP TMDNTKHILN YLMPIIDQVN PELHDFMQSA EVGTIFALSW LITWFGHVLS DFRHVVRLYD FFLAYHPLMP IYFAAVIVLY REQEVLDCDC DMASVHHLLS QIPQDLPYET LISRAGDLFV QFPPSELARE AAAQQQAERT AASTFKDFEL ASAQQRPDMV LRQRFRGLLR PEDRTKDVLT KPRTNRFVKL AVMGLTVALG AAALAVVKSA LEWAPKFQLQ LFP
GTPase-activating protein specific for Rab1 and Rab2 small GTPase families for which it can accelerate the intrinsic GTP hydrolysis rate by more than five orders of magnitude.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TBC1 domain family member 20; TBC1 domain family, member 20
Gene Aliases: C20orf140; TBC1D20; WARBM4
UniProt ID: (Human) Q96BZ9
Entrez Gene ID: (Human) 128637
Molecular Function: GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support