Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TBC1D26 may act as a GTPase-activating protein for Rab family protein(s).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TBC1 domain family member 26; TBC1 domain family, member 26
Gene Aliases: TBC1D26
UniProt ID: (Human) Q86UD7
Entrez Gene ID: (Human) 353149
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support