Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TBC1D28 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TBC1 domain family member 28; TBC1 domain family, member 28
Gene Aliases: TBC1D28
UniProt ID: (Human) Q2M2D7
Entrez Gene ID: (Human) 254272
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support