Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MEQLADVTLR RLLDNEVFDL DPDLQEPSQI TKRDLEARAQ NEFFRAFFRL PRKEKLHAVV DCSLWTPFSR CHTAGRMFAS DSYICFASRE DGCCKIILPL REVVSIEKME
TBC1D8 may act as a GTPase-activating protein for Rab family protein(s).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AD 3; BUB2-like protein 1; TBC1 domain family member 8; TBC1 domain family, member 8 (with GRAM domain); TBC1 domain family, member 8A; Vascular Rab-GAP/TBC-containing protein
Gene Aliases: AD3; GRAMD8; HBLP1; TBC1D8; TBC1D8A; VRP
UniProt ID: (Human) O95759
Entrez Gene ID: (Human) 11138
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support