Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Tubulin-folding protein; involved in the final step of the tubulin folding pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CFC; Tubulin-folding cofactor C; Tubulin-specific chaperone C
Gene Aliases: CFC; TBCC
UniProt ID: (Human) Q15814
Entrez Gene ID: (Human) 6903
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support