Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MESVSCSAAA VRTGDMESQR DLSLVPERLQ RREQERQLEV ERRKQKRQNQ EVEKENSHFF VATFARERAA VEELLERAES VERLEEAASR LQGLQKLIND SVFFLAAYDL RQGQEALARL QAALAERRRG LQPKKRFAFK TRGKDAASST KVDAAPGIPP AVESIQDSPL PKKAEGDLGP SWVCGFSNLE SQVLEKRASE LHQRDVLLTE LSNCTVRLYG NPNTLRLTKA HSCKLLCGPV STSVFLEDCS DCVLAVACQQ LRIHSTKDTR IFLQVTSRAI VEDCSGIQFA PYTWSYPEID KDFESSGLDR SKNNWNDVDD FNWLARDMAS PNWSILPEEE RNIQWD
Tubulin-folding protein; involved in the final step of the tubulin folding pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CFC; Tubulin-folding cofactor C; Tubulin-specific chaperone C
Gene Aliases: 2810055C19Rik; CFC; TBCC
UniProt ID: (Human) Q15814, (Mouse) Q8VCN9
Entrez Gene ID: (Human) 6903, (Rat) 316221, (Mouse) 72726
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support