Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Acts as a regulator of tubulin stability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: catastrophin; E-like; EL; leucine rich repeat containing 35; Leucine-rich repeat-containing protein 35; tubulin folding cofactor E-like; Tubulin-specific chaperone cofactor E-like protein; tubulin-specific chaperone e-like
Gene Aliases: El; LRRC35; TBCEL
UniProt ID: (Human) Q5QJ74
Entrez Gene ID: (Human) 219899
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support