Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Acts as a regulator of tubulin stability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: catastrophin; E-like; EL; leucine rich repeat containing 35; Leucine-rich repeat-containing protein 35; tubulin folding cofactor E-like; Tubulin-specific chaperone cofactor E-like protein; tubulin-specific chaperone e-like
Gene Aliases: El; LRRC35; TBCEL
UniProt ID: (Human) Q5QJ74
Entrez Gene ID: (Human) 219899
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support