Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SNGKYLATC ADDRTIRIWS TKDFLQREHR SMRANVELDH ATLVRFSPDC RAFIVWLANG DTLRVFKMTK REDGGYTFTA TPEDFPKKHK APVIDIGIAN TGKFIMTASS DTTVLIWSLK GQVLSTINTN QMNNTHAAVS PCGRFVASCG FTPDVKVWEV CFGKKGEFQE VVRAFELKGH SAAVHSFAFS NDSRRMASVS KDGTWKLWDT DVEYKKKQDP YLLKTGRFEE AAGAAPCRLA LSPNAQVLAL ASGSSIHLYN TRRGEKEECF ERVHGECIAN LSFDITGRFL ASCGDRAVRL FHNTPGHRAM VEEMQGHLKR ASNESTRQRL QQQLTQAQVT LGGPGAED (100-446 aa encoded by BC012938)
This gene encodes a member of the beta-transducin protein family. Most proteins of the beta-transducin family are involved in regulatory functions. This protein is possibly involved in some intracellular signaling pathway. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP43N024; MGC134739; transducin -like 2; Transducin beta-like protein 2; Williams-Beuren syndrome chromosomal region 13 protein; Williams-Beuren syndrome chromosome region 13; WS beta-transducin repeats protein; WS-betaTRP
Gene Aliases: TBL2; UNQ563/PRO1125; WBSCR13; WS-betaTRP
UniProt ID: (Human) Q9Y4P3
Entrez Gene ID: (Human) 26608
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support