Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDQNNSLPPY AQGLASPQGA MTPGIPIFSP MMPYGTGLTP QPIQNTNSLS ILEEQQRQQQ QQQQQQQQQQ QQQQQQQQQQ QQQQQQQQQQ QQQQAVAAAA VQQSTSQQAT QGTSGQAPQL FHSQTLTTAP LPGTTPLYPS PMTPMTPITP ATPASESSGI VPQLQNIVST VNLGCKLDLK TIALRARNAE YNPKRFAAVI MRIREPRTTA LIFSSGKMVC TGAKSEEQSR LAARKYARVV QKLGFPAKFL DFKIQNMVGS CDVKFPIRLE GLVLTHQQFS SYEPELFPGL IYRMIKPRIV LLIFVSGKVV LTGAKVRAEI YEAFENIYPI LKGFRKTT
RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC117320; MGC126054; MGC126055; RP1-191N21.3; TATA box binding protein; TATA sequence-binding protein; TATA-binding factor; TATA-binding protein; TATA-box binding protein N-terminal domain; TATA-box factor; tata-box-binding; TATA-box-binding protein; TFIID core protein; Transcription initiation factor TFIID TBP subunit
Gene Aliases: GTF2D; GTF2D1; HDL4; SCA17; TBP; TF2D; TFIID
UniProt ID: (Human) P20226, (Mouse) P29037
Entrez Gene ID: (Pig) 110259740, (Human) 6908, (Mouse) 21374, (Rat) 117526
Molecular Function: general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support