Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
TBX15 is a probable transcriptional regulator involved in developmental processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: T-box 14; T-box protein 14; T-box protein 15; T-box transcription factor TBX14; T-box transcription factor TBX15
Gene Aliases: TBX14; TBX15
UniProt ID: (Human) Q96SF7
Entrez Gene ID: (Human) 6913
Molecular Function:
Rel homology transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support