Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: STLLQGTGNG VPATHPHLLS GSSCSSPAFH LGPNTSQLCS LAPADYSACA RSGLTLNRYS TSLAETYNRL TNQAGETFAP PRTPSYVGVS SSTSVNMSMG GTDGDTF
The T-box (Tbx) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The Tbx gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. Tbx genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in Tbx genes are associated with the onset of several human diseases. Our understanding of functional mechanisms of Tbx products has come mainly from the prototypical T/Brachyury, which is a transcription activator. The Tbx genes constitute a family of transcriptional regulatory genes that are implicated in a variety of developmental processes ranging from the formation of germ layers to the organizational patterning of the central nervous system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: T-box protein 18; T-box transcription factor TBX18; TBX14; TBX15
Gene Aliases: CAKUT2; TBX18
UniProt ID: (Human) O95935
Entrez Gene ID: (Human) 9096
Molecular Function:
Rel homology transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support