Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAYHPFHAP RPADFPMSAF LAAAQPSFFP ALALPPGALA KPLPDPGLAG AAAAAAAAAA AAEAGLHVSA LGPHPPAAHL RSLKSLEPED EVEDDPKVTL EAKELWDQFH KLGTEMVITK SGRRMFPPFK VRVSGLDKKA KYILLMDIVA ADDCRYKFHN SRWMVAGKAD PEMPKRMYIH PDSPATGEQW MAKPVAFHKL KLTNNISDKH GFTILNSMHK YQPRFHIVRA NDILKLPYST FRTYVFPETD FIAVTAYQND KITQLKIDNN PFAKGFRDTG NGRREKRKQL TLPSLRLYEE HCKPERDGAE SDASSCDPPP AREPPTSPGA APSPLRLHRA RAEEKSCAAD (1-349 aa encoded by BC052566)
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene product is the human homolog of mouse Tbx2, and shares strong sequence similarity with Drosophila omb protein. Expression studies indicate that this gene may have a potential role in tumorigenesis as an immortalizing agent. Transcript heterogeneity due to alternative polyadenylation has been noted for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10169; T-box protein 2; T-box transcription factor TBX2
Gene Aliases: TBX2
UniProt ID: (Human) Q13207
Entrez Gene ID: (Human) 6909
Molecular Function:
Rel homology transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support