Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: DQYLQAPNST NQMLYGLQSP GNIFLPNSIT PEALSCSFHP SYDFYRYNFS MPSRLISGSN HLKVNDDSQV SFGEGKCNHV HWYPAINHY
The T-box (TBX) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The TBX gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. TBX genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in TBX genes are associated with the onset of several human diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: T-box protein 22; T-box transcription factor TBX22
Gene Aliases: ABERS; CLPA; CPX; dJ795G23.1; TBOX22; TBX22; TBXX
UniProt ID: (Human) Q9Y458
Entrez Gene ID: (Human) 50945
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support