Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TTTSEEPCR DKGSPAVKAH LFAAERPRDS GRLDKASPDS RHSPATISSS TRGLGAEERR SPVREGTAPA KVEEARALPG KEAFAPLTVQ TDAAAAHLAQ GPLPGLGFAP GLAGQQFFNG HPLFLHPSQF AMGGAFSSMA AAGMGPLLAT VSGASTGVSG LDSTAMASAA AAQGLSGASA ATLPFHLQQH VLASQGLAMS PFGSLFPYPY TYMAAAAAAS SAAASSSVHR HPFLNLNTMR PRLRYSPYSI PVPVPDGSSL LTTALPSMAA AAGPLDGKVA ALAASPASVA VDSGSELNSR SSTLSSSSMS LSPKLCAEKE AATSELQSIQ RLVSGLEAKP DRSRSASP (397-743 aa encoded by BC025258)
T-box 3 (TBX3) is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. The Tbx3 protein is a transcriptional repressor that binds the canonical Brachychury bindig sites, and is thought to play a role in the anterior/posterior axis of the tetrapod forelimb. Tbx3 is required for normal mammary development and is also implicated in tumor development. Tbx3 expression also increases during osteoblast differentiation, and may function as a determinant of osteoblast cell numbers. Mutations in T-box 3 cause ulnar-mammary syndrome (MIM 181450), affecting limb, apocrine gland, tooth, hair, and genital development. Alternative splicing results in three transcript variants encoding different isoforms; however, the nature of one full length variant has not been determined. Subsets of human breast cancer cell lines overexpress TBX36. in vitro, Tbx3 overexpression in mouse embryo fibroblasts leads to immortalization of cells. Truncated forms of Tbx3 are increased in plasma samples.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bladder cancer related protein XHL; T-box protein 3; T-box transcription factor TBX3
Gene Aliases: D5Ertd189e; TBX3; TBX3-ISO; UMS; XHL
UniProt ID: (Human) O15119, (Mouse) P70324
Entrez Gene ID: (Human) 6926, (Mouse) 21386
Molecular Function:
Rel homology transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support