Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MADADEGFGL AHTPLEPDAK DLPCDSKPES ALGAPSKSPS SPQAAFTQQG MEGIKVFLHE RELWLKFHEV GTEMIITKAG RRMFPSYKVK VTGLNPKTKY ILLMDIVPAD DHRYKFADNK WSVTGKAEPA MPGRLYVHPD SPATGAHWMR QLVSFQKLKL TNNHLDPFGH IILNSMHKYQ PRLHIVKADE NNGFGSKNTA FCTHVFPETA FIAVTSYQNH KITQLKIENN PFAKGFRGSD DMELHRMSRM QSKEYPVVPR STVRQKVASN HSPFSSESRA LSTSSNLGSQ YQCENGVSGP SQDLLPPPNP YPLPQEHSQI YHCTKRKEEE CSTTDHPYKK PYMETSPSEE DSFYRSSYPQ QQGLGASYRT ESAQRQACMY ASSAPPSEPV PSLEDISCNT WPSMPSYSSC TVTTVQPMDR LPYQHFSAHF TSGPLVPRLA GMANHGSPQL GEGMFQHQTS VAHQPVVRQC GPQTGLQSPG TLQPPEFLYS HGVPRTLSPH QYHSVHGVGM VPEWSDNS
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene is closely linked to related family member T-box 3 (ulnar mammary syndrome) on human chromosome 12. The encoded protein may play a role in heart development and specification of limb identity. Mutations in this gene have been associated with Holt-Oram syndrome, a developmental disorder affecting the heart and upper limbs. Several transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: T-box protein 5; T-box transcription factor TBX5
Gene Aliases: HOS; TBX5
UniProt ID: (Human) Q99593, (Mouse) P70326
Entrez Gene ID: (Human) 6910, (Mouse) 21388
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support