Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total TAS2R.
The thromboxane A2 receptor (TBXA2R) is a member of the Prostanoid Receptor subfamily. TBXA2R performs an essential role in hemostasis by interacting with thromboxane A2 to induce platelet aggregation. Thromboxane A2 has been implicated in myocardial infarct, stroke, and asthma. Additionally, thromboxane-prostanoid receptors have been implicated in the pathogenesis of cardiovascular diseases. Changes in TBXA2R function have also been linked to a hereditary bleeding disorder.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Prostanoid TP receptor; Thromboxane A2 receptor; thromboxane prostanoid; Thromboxane receptor; TXA2 receptor; TXA2-R; TXR2
Gene Aliases: BDPLT13; TBXA2R; TP; TXA2-R
UniProt ID: (Human) P21731, (Mouse) P30987, (Rat) P34978
Entrez Gene ID: (Human) 6915, (Mouse) 21390, (Rat) 24816
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support