Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Additional Information: This antibody is expected to recognize the reported isoform beta (NP_963998.2) only.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Receptor for thromboxane A2 (TXA2), a potent stimulator of platelet aggregation. The activity of this receptor is mediated by a G-protein that activates a phosphatidylinositol-calcium second messenger system. In the kidney, the binding of TXA2 to glomerular TP receptors causes intense vasoconstriction. Activates phospholipase C. This target is specific to isoform beta, endothelial receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Prostanoid TP receptor; Thromboxane A2 receptor; TXA2-R
Gene Aliases: BDPLT13; TBXA2R; TXA2-R
UniProt ID: (Human) P21731
Entrez Gene ID: (Human) 6915
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support