Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Control: 293 cell lysates.
TC10 is a member of the RAS superfamily of small GTP-binding proteins (see HRAS, MIM 190020) involved in insulin-stimulated glucose uptake.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 42; ras homolog gene family, member Q; Ras-like protein family member 7A; Ras-like protein TC10; RAS-like, family 7, member A; Rho-related GTP-binding protein RhoQ
Gene Aliases: ARHQ; HEL-S-42; RASL7A; RHOQ; TC10; TC10A
UniProt ID: (Human) P17081
Entrez Gene ID: (Human) 23433
Molecular Function:
small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support