Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
TCEAL8 is a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. This gene encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome. Alternative splicing results in multiple transcript variants encoding a single isoform.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC45400; TCEA-like protein 8; transcription elongation factor A (SII)-like 8; Transcription elongation factor A protein-like 8; Transcription elongation factor S-II protein-like 8; Up-regulated in nephrectomized rat kidney #1
Gene Aliases: 3930402F23Rik; TCEAL8; UR-NR#1; WEX3
UniProt ID: (Human) Q8IYN2, (Rat) Q6I7R5, (Mouse) Q9CZY2
Entrez Gene ID: (Human) 90843, (Rat) 367909, (Mouse) 66684
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support