Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: ETDVVRAVYR VLKNCPSVAL KKKAKCLLSK WKAVYKQTHS KARNSPKLFP VRGNKEENSG PSHDPSQNET LGICSSNSLS SQDVAKLSEM IVPENRAIQL KPKEEHFGDG DPESTGK
Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 53%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TFIIS central domain-containing protein 1; TFS2-M domain-containing protein 1; transcription elongation factor A (SII) N-terminal and central domain containing; Transcription elongation factor A N-terminal and central domain-containing protein
Gene Aliases: TCEANC
UniProt ID: (Human) Q8N8B7
Entrez Gene ID: (Human) 170082
Molecular Function:
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support