Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
TCEANC2 (Transcription Elongation Factor A N-Terminal And Central Domain Containing 2) is a Protein Coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: transcription elongation factor A (SII) N-terminal and central domain containing 2; Transcription elongation factor A N-terminal and central domain-containing protein 2
Gene Aliases: C1orf83; TCEANC2
UniProt ID: (Human) Q96MN5
Entrez Gene ID: (Human) 127428
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support