Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
TCEB1 is known as protein elongin C, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EloC; Elongin 15 kDa subunit; elongin C; Elongin-C; RNA polymerase II transcription factor SIII subunit C; SIII p15; Stromal membrane-associated protein SMAP1B homolog; transcription elongation factor B (SIII), polypeptide 1 (15kDa, elongin C); Transcription elongation factor B polypeptide 1
Gene Aliases: 2610043E24Rik; 2610301I15Rik; AA407206; AI987979; AW049146; ELOC; SIII; TCEB1
UniProt ID: (Human) Q15369, (Mouse) P83940
Entrez Gene ID: (Human) 6921, (Mouse) 67923
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support