Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: RHKTTIFTDA KESSTVFELK RIVEGILKRP PDEQRLYKDD QLLDDGKTLG ECGFTSQTAR PQAPATVGLA FRADDTFEAL CIEPFSSPPE LPDVMKPQDS GSSANEQAVQ
SIII, also known as elongin, is a general transcription elongation factor that increases the RNA polymerase II transcription elongation past template-encoded arresting sites. Subunit A is transcriptionally active and its transcription activity is strongly enhanced by binding to the dimeric complex of the SIII regulatory subunits B and C (elongin BC complex).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EloB; Elongin 18 kDa subunit; elongin B; elongin, 18-kD subunit; Elongin-B; RNA polymerase II transcription factor SIII p18 subunit; RNA polymerase II transcription factor SIII subunit B; SIII p18; TCEB 2; transcription elongation factor B (SIII) polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B); Transcription elongation factor B polypeptide 2
Gene Aliases: ELOB; SIII; TCEB2
UniProt ID: (Human) Q15370, (Rat) P62870
Entrez Gene ID: (Human) 6923, (Rat) 81807
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support