Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
TCF7L1 is one of TCF/LEF transcription factors that are mediators of the Wnt signaling pathway and are antagonized by the TGF-beta signaling pathway. It binds to DNA and acts as a repressor in the absence of CTNNB1, and as an activator in its presence. It's necessary for the terminal differentiation of epidermal cells, the formation of keratohyalin granules and the development of the barrier function of the epidermis. This antibody raised against 1-200aa of human TCF7L1 can also react with TCF7L2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HMG box transcription factor 3; HMG box transcription factor 4; hTCF-4; mTCF-4; T-cell factor 3; T-cell factor 4; T-cell factor-4 variant A; T-cell factor-4 variant B; T-cell factor-4 variant C; T-cell factor-4 variant D; T-cell factor-4 variant E; T-cell factor-4 variant F; T-cell factor-4 variant G; T-cell factor-4 variant H; T-cell factor-4 variant I; T-cell factor-4 variant J; T-cell factor-4 variant K; T-cell factor-4 variant L; T-cell factor-4 variant M; T-cell factor-4 variant X2; T-cell-specific transcription factor 4; TCF-3; TCF7L1/2; transcription factor 3; Transcription factor 7-like 1; transcription factor 7-like 1 (T-cell specific, HMG box); transcription factor 7-like 1 (T-cell specific, HMG-box); Transcription factor 7-like 2; transcription factor 7-like 2 (T-cell specific, HMG box); transcription factor 7-like 2 (T-cell specific, HMG-box); transcription factor 7-like 2, T-cell specific, HMG-box
Gene Aliases: bHLHb21; mTcf-4B; mTcf-4E; TCF-3; TCF-4; TCF3; TCF4; TCF4B; TCF4E; TCF7L1; TCF7L2
UniProt ID: (Human) Q9HCS4, (Human) Q9NQB0, (Mouse) Q9Z1J1, (Mouse) Q924A0
Entrez Gene ID: (Human) 83439, (Human) 6934, (Mouse) 21415, (Mouse) 21416
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support