Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs, less than 5% cross-reactivity with recombinant human TCL1A is observed.
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
TCL1B (T Cell Leukemia/Lymphoma 1B) is a Protein Coding gene. Diseases associated with TCL1B include T-Cell Leukemia and Lymphoma. Among its related pathways are PI3K-Akt signaling pathway. An important paralog of this gene is MTCP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Oncogene TCL-1B; Oncogene TCL1B; SYN-1; Syncytiotrophoblast-specific protein; T-cell leukemia/lymphoma protein 1B; T-cell lymphoma/leukemia 1B; TCL1/ MTCP1-like 1; TCL1/MTCP1-like protein 1
Gene Aliases: SYN-1; TCL1; TCL1B; TML1
UniProt ID: (Human) O95988
Entrez Gene ID: (Human) 9623
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support