Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MRQSHQLPLV GLLLFSFIPS QLCEICEVSE ENYIRLKPLL NTMIQSNYNR GTSAVNVVLS LKLVGIQIQT LMQKMIQQIK YNVKSRLSDV SSGELALIIL ALGVCRNAEE NLIYDYHLID KLENKFQAEI ENMEAHNGTP LTNYYQLSLD VLALCLFNGN YSTAEVVNHF TPENKNYYFG SQFSVDTGAM AVLALTCVKK SLINGQIKAD EGSLKNISIY TKSLVEKILS EKKENGLIGN TFSTGEAMQA LFVSSDYYNE NDWNCRQTLN TVLTEISQGA FSNPNAAAQV LPALMGKTFL DINKDSSCVP ASGNFNISAD EPITVTPPDS QSYISVDYSV RINETYFTNV AVLNGSVFLS VMEKAQKMND TIFGFTMEER SWGPYITCIQ GLCANNNDRT YWELLSGGEP LSQGAGSYVV RNG
Transcobalamin I (TCI) and Transcobalamin II (TCII) are secreted proteins belonging to the eukaryotic cobalamin transport proteins family and also to the vitamin B12-binding protein family. The genes encoding these proteins map to chromosome 11q11-q12 and 22q12.2, respectively. Transcobalamin I is a constituent of secondary granules in neutrophils, while Transcobalamin II binds cobalamin and mediates its transport into cells. These plasma proteins are expressed in various tissues and secretions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: haptocorin; Haptocorrin; HC; Protein R; TC I; TC-1; Transcobalamin I; transcobalamin I (vitamin B12 binding protein, R binder family); Transcobalamin-1
Gene Aliases: HC; TC-1; TC1; TCI; TCN1
UniProt ID: (Human) P20061
Entrez Gene ID: (Human) 6947
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support