Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ESSRISDGKK QEGPATQVSK KNPASLPLTQ AALKVLAQKA SEAQPPVART QPSSGVDSAV GTLPATSPQS TSVQAKGTNK LRKPK
Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 32%.
Treacle is a serine/alanine-rich 1,411 amino acid nuclear phosphoprotein that interacts with upstream binding factor (UBF) and affects transcription of the ribosomal DNA gene. Treacle is also involved in early embryonic development, particularly in the craniofacial complex, and may play a role in nucleolarcytoplasmic transport. The Treacle protein contains three domains with unique N- and C-termini and a large central repeat domain. Mutations in TCOF1, the gene that encodes for Treacle, cause Treacher Collins-Franceschetti syndrome (TCS), a disorder characterized by defects in craniofacial development. Symptoms of TCS include conductive hearing loss, hypoplasia of the mandible and maxilla, downward sloping palpebral fissures and cleft palate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: nucleolar trafficking phosphoprotein; Treacher Collins syndrome protein; Treacher Collins-Franceschetti syndrome 1; Treacle protein
Gene Aliases: MFD1; TCOF1; TCS; TCS1; treacle
UniProt ID: (Human) Q13428
Entrez Gene ID: (Human) 6949
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support