Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 14
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
The chaperonin containing TCP1 (MIM 186980) complex (CCT), also called the TCP1 ring complex, consists of 2 back-to-back rings, each containing 8 unique but homologous subunits, such as CCT4. CCT assists the folding of newly translated polypeptide substrates through multiple rounds of ATP-driven release and rebinding of partially folded intermediate forms. Substrates of CCT include the cytoskeletal proteins actin (see MIM 102560) and tubulin (see MIM 191130), as well as alpha-transducin (MIM 139330) (Won et al., 1998 [PubMed 9819444]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A45; CCT-delta; chaperonin containing t-complex polypeptide 1, delta subunit; chaperonin containing TCP1, subunit 4 (delta); chaperonin subunit 4 (delta); Stimulator of TAR RNA-binding; T complex protein 1, delta; T-complex protein 1 subunit delta; TCP-1 delta; TCP-1-delta
Gene Aliases: 2610204B21Rik; A45; C78323; CCT-DELTA; CCT4; CCTD; SRB
UniProt ID: (Human) P50991, (Mouse) P80315, (Rat) Q7TPB1
Entrez Gene ID: (Human) 10575, (Mouse) 12464, (Rat) 29374
Molecular Function:
chaperonin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support