Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLAGQLEAR DPKEGTHPED PCPGAGAVME KTAVAAEVLT EDCNTGEMPP LQQQIIRLHQ ELGRQKSLWA DVHGKLRSHI DALREQNMEL REKLRALQLQ RWKARKKSAA SPHAGQESHT LALEPAFGKI SPLSADEETI PKYAGHKNQS ATLLGQRSSS NNSAPPKPMS LKIERISSWK TPPQENRDKN LSRRRQDRRA TPTGRPTPCA ERRGGV (1-215 aa encoded by B C033855)
TCP10L is a human leucine zipper protein with transcription inhibition activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: t-complex 10 (a murine tcp homolog)-like; T-complex 10A-2; T-complex protein 10A homolog 1; T-complex protein 10A-1; T-complex protein 10A-2; TCP10-like
Gene Aliases: C21orf77; PRED77; TCP10A-2; TCP10L
UniProt ID: (Human) Q8TDR4
Entrez Gene ID: (Human) 140290
Molecular Function:
cytoskeletal protein
microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support