Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LKVLYLSKY VLNMMALLCA PVRDEAVQKL ENITDPVWLL RGIFQVLGRM KMDMVNYTIQ SLQPHLQEHS IQYERAKFQE LLNKQPSLLN HTTKWLTQAA GDLTMSPPTC PDTSDSSSVA GPSPNEAANN PEPLSPTMVL CQGFLNLLLW DLENEEFPET LLMDRTRLQE LKSQLHQLTV MASVLLVASS FSGSVLFGSP QFVDKLKRIT KSLLEDFHSR PEEAILTVSE QVSQEIHQSL KNMGLVALSS DNTASLMGQL QNIAKKENCV CSVIDQRIHL FLKCCLVLGV QRSLLDLPGG LTLIEAELAE LGQKFVNLTH HNQQVFGPYY TEILKTLISP AQALETKVES V (92-441 aa encoded by BC048418)
Plays a role in the process of sperm capacitation and acrosome reactions. Probable receptor for the putative fertilization-promoting peptide (FPP) at the sperm membrane that may modulate the activity of the adenylyl cyclase cAMP pathway. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fertilization-promoting peptide receptor; t-complex 11; t-complex 11 (a murine tcp homolog); t-complex 11 homolog; t-complex 11, testis-specific; T-complex protein 11; T-complex protein 11 homolog; testis secretory sperm-binding protein Li 222n; testis-specific protein PBS13
Gene Aliases: D17Ken1; D6S230E; FPPR; Tcp-11; TCP11
UniProt ID: (Human) Q8WWU5, (Rat) Q5XI00
Entrez Gene ID: (Human) 6954, (Rat) 309641, (Mouse) 21463
Molecular Function:
microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support