Product References
Ibuprofen supports macrophage differentiation, T cell recruitment, and tumor suppression in a model of postpartum breast cancer.
Journal for immunotherapy of cancer
Pennock ND,Martinson HA,Guo Q,Betts CB,Jindal S,Tsujikawa T,Coussens LM,Borges VF,Schedin P
17-5808 was used in Functional assays to implicate beneficial prophylactic use of ibuprofen during the pro-tumorigenic window of mammary gland involution.
Mon Oct 01 00:00:00 EDT 2018
PDL2+ CD11b+ dermal dendritic cells capture topical antigen through hair follicles to prime LAP+ Tregs.
Nature communications
Tordesillas L,Lozano-Ojalvo D,Dunkin D,Mondoulet L,Agudo J,Merad M,Sampson HA,Berin MC
17-5808 was used in Flow cytometry/Cell sorting to show how topical antigen is acquired and presented by dendritic cells in the skin.
Fri Dec 07 00:00:00 EST 2018
Pneumococcal Surface Protein A Plays a Major Role in Streptococcus pneumoniae-Induced Immunosuppression.
Journal of immunology (Baltimore, Md. : 1950)
Pujanauski L,Colino J,Flora M,Torres RM,Tuomanen E,Snapper CM
17580880 was used in flow cytometry to elucidate the immunosuppressive effect of intact, inactivated Streptococcus pneumoniae treatment
Sun May 01 00:00:00 EDT 2016
RGMb is a novel binding partner for PD-L2 and its engagement with PD-L2 promotes respiratory tolerance.
The Journal of experimental medicine
Xiao Y,Yu S,Zhu B,Bedoret D,Bu X,Francisco LM,Hua P,Duke-Cohan JS,Umetsu DT,Sharpe AH,DeKruyff RH,Freeman GJ
17-5808 was used in Flow cytometry/Cell sorting to show that programmed cell death ligand 2 (PD-L2) binds to repulsive guidance molecule b (RGMb) to promote respiratory tolerance.
Mon May 05 00:00:00 EDT 2014
C5a receptor signalling in dendritic cells controls the development of maladaptive Th2 and Th17 immunity in experimental allergic asthma.
Mucosal immunology
Schmudde I,Ströver HA,Vollbrandt T,König P,Karsten CM,Laumonnier Y,Köhl J
17-5808 was used in Flow cytometry/Cell sorting to uncover roles for C5aR in Th17 differentiation, T-cell survival, and DC-suppressor population differentiation, controlling Th2 immunity in experimental allergic asthma.
Mon Jul 01 00:00:00 EDT 2013
STAT6 controls the number of regulatory T cells in vivo, thereby regulating allergic lung inflammation.
Journal of immunology (Baltimore, Md. : 1950)
Dorsey NJ,Chapoval SP,Smith EP,Skupsky J,Scott DW,Keegan AD
17-5808 was used in Flow cytometry/Cell sorting to study the interactions between STAT6 and Tregs during in vivo allergic airway inflammation.
Thu Aug 15 00:00:00 EDT 2013
Protein-bound polysaccharide activates dendritic cells and enhances OVA-specific T cell response as vaccine adjuvant.
Immunobiology
Engel AL,Sun GC,Gad E,Rastetter LR,Strobe K,Yang Y,Dang Y,Disis ML,Lu H
17-5808 was used in Flow cytometry/Cell sorting to evaluate the effect of protein-bound polysaccharide-K on dendritic cell activation and its potential as a vaccine adjuvant.
Sun Dec 01 00:00:00 EST 2013
S-nitrosoglutathione reductase inhibition regulates allergen-induced lung inflammation and airway hyperreactivity.
PloS one
Ferrini ME,Simons BJ,Bassett DJ,Bradley MO,Roberts K,Jaffar Z
17-5808 was used in Flow cytometry/Cell sorting to study the effects of SPL-334 on airway hyperreactivity, allergen-specific T cell and eosinophil accumulation, and mucus production in the lungs in response to allergen inhalation.
Wed Mar 05 00:00:00 EST 2014
Rapid in vivo conversion of effector T cells into Th2 cells during helminth infection.
Journal of immunology (Baltimore, Md. : 1950)
Panzer M,Sitte S,Wirth S,Drexler I,Sparwasser T,Voehringer D
17-5808 was used in Flow cytometry/Cell sorting to determine whether different classes of T cells can be converted into IL-4-expressing Th2 cells in vivo by infection of mice with gastrointestinal helminth.
Sun Jan 15 00:00:00 EST 2012
TLR2-dependent modulation of dendritic cells by LT-IIa-B5, a novel mucosal adjuvant derived from a type II heat-labile enterotoxin.
Journal of leukocyte biology
Lee CH,Masso-Welch P,Hajishengallis G,Connell TD
17-5808-80 was used in Flow Cytometry to demonstrate that LT-IIa-B(5) enhances the mucosal immune response by modulating the activities of dendritic cells.
Tue Nov 01 00:00:00 EDT 2011
Prostaglandin I₂promotes the development of IL-17-producing γδ T cells that associate with the epithelium during allergic lung inflammation.
Journal of immunology (Baltimore, Md. : 1950)
Jaffar Z,Ferrini ME,Shaw PK,FitzGerald GA,Roberts K
17-5808 was used in Flow cytometry/Cell sorting to show that allergic airway inflammation was associated with an increase in IL-17-producing gd T-cells associated with airway epithelium.
Tue Nov 15 00:00:00 EST 2011
Natural Foxp3(+) regulatory T cells inhibit Th2 polarization but are biased toward suppression of Th17-driven lung inflammation.
Journal of leukocyte biology
Girtsman T,Jaffar Z,Ferrini M,Shaw P,Roberts K
17-5808 was used in Flow cytometry/Cell sorting to study the role of nTregs in suppressing the development of lung inflammatory responses, elicited by differentiated CD4(+) T cells following antigen inhalation.
Wed Sep 01 00:00:00 EDT 2010
Costimulator B7-DC attenuates strong Th2 responses induced by Nippostrongylus brasiliensis.
Journal of immunology (Baltimore, Md. : 1950)
Ishiwata K,Watanabe N,Guo M,Tomihara K,Brumlik MJ,Yagita H,Pardoll D,Chen L,Shin T
17-5808 was used in Flow cytometry/Cell sorting to demonstrate that the inhibition of Th2 responses by B7-DC occurs independently of programmed death 1 but conceivably acts through an as yet unknown alternative receptor that enhances Th1 responses.
Mon Feb 15 00:00:00 EST 2010
Tracking epitope-specific T cells.
Nature protocols
Moon JJ,Chu HH,Hataye J,Pagán AJ,Pepper M,McLachlan JB,Zell T,Jenkins MK
Published figure using TCR DO11.10 monoclonal antibody (Product # 17-5808-80) in Flow Cytometry
Tue Jun 30 00:00:00 EDT 2009