Product References
Longitudinal profiling of respiratory and systemic immune responses reveals myeloid cell-driven lung inflammation in severe COVID-19.
Immunity
Szabo PA,Dogra P,Gray JI,Wells SB,Connors TJ,Weisberg SP,Krupska I,Matsumoto R,Poon MML,Idzikowski E,Morris SE,Pasin C,Yates AJ,Ku A,Chait M,Davis-Porada J,Guo XV,Zhou J,Steinle M,Mackay S,Saqi A,Baldwin MR,Sims PA,Farber DL
Published figure using TCR gamma/delta monoclonal antibody (Product # 11-9959-42) in Flow Cytometry
Tue Apr 13 00:00:00 EDT 2021
γδT Cells Are Required for CD8+ T Cell Response to Vaccinia Viral Infection.
Frontiers in immunology
Dai R,Huang X,Yang Y
11-9959-42 was used in Flow cytometry/Cell sorting to illustrate a critical role for γδT cells in the regulation of adaptive T cell response to viral infection.
Fri Nov 26 00:00:00 EST 2021
SPARC regulation of PMN clearance protects from pristane-induced lupus and rheumatoid arthritis.
iScience
Sangaletti S,Botti L,Gulino A,Lecis D,Bassani B,Portararo P,Milani M,Cancila V,De Cecco L,Dugo M,Tripodo C,Colombo MP
Published figure using TCR gamma/delta monoclonal antibody (Product # 11-9959-42) in Flow Cytometry
Fri Jun 25 00:00:00 EDT 2021
Systems Biology Methods Applied to Blood and Tissue for a Comprehensive Analysis of Immune Response to Hepatitis B Vaccine in Adults.
Frontiers in immunology
Ben-Othman R,Cai B,Liu AC,Varankovich N,He D,Blimkie TM,Lee AH,Gill EE,Novotny M,Aevermann B,Drissler S,Shannon CP,McCann S,Marty K,Bjornson G,Edgar RD,Lin DTS,Gladish N,Maclsaac J,Amenyogbe N,Chan Q,Llibre A,Collin J,Landais E,Le K,Reiss SM,Koff WC,Havenar-Daughton C,Heran M,Sangha B,Walt D,Krajden M,Crotty S,Sok D,Briney B,Burton DR,Duffy D,Foster LJ,Mohn WW,Kobor MS,Tebbutt SJ,Brinkman RR,Scheuermann RH,Hancock REW,Kollmann TR,Sadarangani M
Published figure using TCR gamma/delta monoclonal antibody (Product # 11-9959-42) in Flow Cytometry
Mon Jun 21 00:00:00 EDT 2021
Biological Therapy in Inflammatory Bowel Disease Patients Partly Restores Intestinal Innate Lymphoid Cell Subtype Equilibrium.
Frontiers in immunology
Creyns B,Jacobs I,Verstockt B,Cremer J,Ballet V,Vandecasteele R,Vanuytsel T,Ferrante M,Vermeire S,Van Assche G,Ceuppens JL,Breynaert C
11-9959 was used in Flow cytometry/Cell sorting to study the impact of biological treatment on innate lymphoid cells in Crohn disease and ulcerative colitis.
Mon Apr 12 00:00:00 EDT 2021
Th17 reprogramming of T cells in systemic juvenile idiopathic arthritis.
JCI insight
Henderson LA,Hoyt KJ,Lee PY,Rao DA,Jonsson AH,Nguyen JP,Rutherford K,Julé AM,Charbonnier LM,Case S,Chang MH,Cohen EM,Dedeoglu F,Fuhlbrigge RC,Halyabar O,Hazen MM,Janssen E,Kim S,Lo J,Lo MS,Meidan E,Son MBF,Sundel RP,Stoll ML,Nusbaum C,Lederer JA,Chatila TA,Nigrovic PA
11-9959 was used in Flow cytometry/Cell sorting to show how systemic juvenile idiopathic arthritis (sJIA) transitions from acute to chronic through the evolving Th17 polarization that begins in Tregs and progresses to T effector cells (Teffs).
Thu Mar 26 00:00:00 EDT 2020
CD62L Is a Functional and Phenotypic Marker for Circulating Innate Lymphoid Cell Precursors.
Journal of immunology (Baltimore, Md. : 1950)
Bar-Ephraim YE,Koning JJ,Burniol Ruiz E,Konijn T,Mourits VP,Lakeman KA,Boon L,Bögels M,van Maanen JP,Den Haan JMM,van Egmond M,Bouma G,Reijmers RM,Mebius RE
11-9959-42 was used in Flow Cytometry to demonstrate that CD62L not only serves as a functional marker of ILCP, but has potential to be used in the clinic as a diagnostic marker in inflammatory disorders.
Tue Jan 01 00:00:00 EST 2019
Genetic Architecture of Adaptive Immune System Identifies Key Immune Regulators.
Cell reports
Lagou V,Garcia-Perez JE,Smets I,Van Horebeek L,Vandebergh M,Chen L,Mallants K,Prezzemolo T,Hilven K,Humblet-Baron S,Moisse M,Van Damme P,Boeckxstaens G,Bowness P,Dubois B,Dooley J,Liston A,Goris A
11-9959-42 was used in Flow Cytometry to identify eight genome-wide significant associations explaining 6%-20% of variance in immune system regulation.
Tue Oct 16 00:00:00 EDT 2018
High Prevalence of MERS-CoV Infection in Camel Workers in Saudi Arabia.
mBio
Alshukairi AN,Zheng J,Zhao J,Nehdi A,Baharoon SA,Layqah L,Bokhari A,Al Johani SM,Samman N,Boudjelal M,Ten Eyck P,Al-Mozaini MA,Zhao J,Perlman S,Alagaili AN
11-9959 was used in Flow cytometry/Cell sorting to show that approximately 50% of camel workers in the Kingdom of Saudi Arabia and 0% of controls were previously infected.
Tue Oct 30 00:00:00 EDT 2018
Systemic Human ILC Precursors Provide a Substrate for Tissue ILC Differentiation.
Cell
Lim AI,Li Y,Lopez-Lastra S,Stadhouders R,Paul F,Casrouge A,Serafini N,Puel A,Bustamante J,Surace L,Masse-Ranson G,David E,Strick-Marchand H,Le Bourhis L,Cocchi R,Topazio D,Graziano P,Muscarella LA,Rogge L,Norel X,Sallenave JM,Allez M,Graf T,Hendriks RW,Casanova JL,Amit I,Yssel H,Di Santo JP
11-9959 was used in Flow cytometry/Cell sorting to identify circulating and tissue ILC precursors in humans that fail to express the transcription factors and cytokine outputs of mature ILCs but have these signature loci in an epigenetically poised configuration.
Thu Mar 09 00:00:00 EST 2017
Early changes within the lymphocyte population are associated with the development of multiple organ dysfunction syndrome in trauma patients.
Critical care (London, England)
Manson J,Cole E,De'Ath HD,Vulliamy P,Meier U,Pennington D,Brohi K
11-9959 was used in Flow cytometry/Cell sorting to investigate the relationship between early lymphocyte responses and the development of MODS.
Tue Jun 07 00:00:00 EDT 2016
Evidence for Resident Memory T Cells in Rasmussen Encephalitis.
Frontiers in immunology
Owens GC,Chang JW,Huynh MN,Chirwa T,Vinters HV,Mathern GW
11-9959 was used in Flow cytometry/Cell sorting to demonstrate that a cellular immune response to an as yet unidentified antigen(s) occurs at an early stage of Rasmussen encephalitis.
Fri Mar 04 00:00:00 EST 2016
Neutrophils in antiretroviral therapy-controlled HIV demonstrate hyperactivation associated with a specific IL-17/IL-22 environment.
The Journal of allergy and clinical immunology
Campillo-Gimenez L,Casulli S,Dudoit Y,Seang S,Carcelain G,Lambert-Niclot S,Appay V,Autran B,Tubiana R,Elbim C
11-9959 was used in Flow cytometry/Cell sorting to analyse polymorphonuclear leukocyte function in ART-treated and controlled HIV-infected patients with and without inflammatory diseases.
Sat Nov 01 00:00:00 EDT 2014
Differentiation of human peripheral blood Vδ1+ T cells expressing the natural cytotoxicity receptor NKp30 for recognition of lymphoid leukemia cells.
Blood
Correia DV,Fogli M,Hudspeth K,da Silva MG,Mavilio D,Silva-Santos B
11-9959 was used in Flow cytometry/Cell sorting to implicate NKp30(+) Vδ1(+) T-cells as a novel, inducible and specialized killer lymphocyte population with potential for human cancer immunotherapy.
Thu Jul 28 00:00:00 EDT 2011
Gamma interferon secretion by human Vgamma2Vdelta2 T cells after stimulation with antibody against the T-cell receptor plus the Toll-Like receptor 2 agonist Pam3Cys.
Infection and immunity
Deetz CO,Hebbeler AM,Propp NA,Cairo C,Tikhonov I,Pauza CD
Tue Aug 01 00:00:00 EDT 2006