Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Keep as concentrated solution.
Predicted reactivity: Human (99%), Mouse (88%), Rat (87%), Bovine (91%).
Positive Control: human TDG-transfected 293T cells.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene belongs to the TDG/mug DNA glycosylase family. Thymine-DNA glycosylase removes thymine moieties from G/T mismatches by hydrolyzing the carbon-nitrogen bond between the sugar-phosphate backbone of DNA and the mis paired thymine. With lower activity, this enzyme also removes thymine from C/T and T/T mispairings. TDG can also remove uracil and 5-bromouracil from mispairings with guanine. This enzyme plays a central role in cellular defense against genetic mutation caused by the spontaneous deamination of 5-methylcytosine and cytosine. This gene may have a pseudogene in the p arm of chromosome 12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-JUN leucine zipper interactive protein JZA-3; G/T mismatch-specific thymine DNA glycosylase; hTDG; Thymine-DNA glycosylase
Gene Aliases: E130317C12Rik; hTDG; JZA-3; Jza1; TDG
UniProt ID: (Human) Q13569, (Mouse) P56581
Entrez Gene ID: (Human) 6996, (Rat) 114521, (Mouse) 21665
Molecular Function: DNA metabolism protein DNA glycosylase nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support