Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSTERTSWTS LSTIQKIALG FGIPASATVA YILYRRYRES REERLTFVGE DDIEIEMRVP QEAVKLIIGR QGANIKQLRK QTGARIDVDT EDVGDERVLL ISGFPVQVCK AKAAIHQILT ENTPVSEQLS VPQRSVGRII GRGGETIRSI CKASGAKITC DKESEGTLLL SRLIKISGTQ KEVAAAKHLI LEKVSEDEEL RKRIAHSAET RVPRKQPISV RREDMTEPGG AGEPALWKNT SSSMEPTAPL VTPPPKAGGD MAVVVSKEGS WEKPSDDSFQ KSEAQAIPEM PMFEIPSPDF SFHADEYLEV YVSASEHPNH FWIQIVGSRS LQLDKLVNEM TQHYENSVPE DLTVHVGDIV AAPLPTNGSW YRAR
TDRKH is a protein of unknown function containing one tudor domain, and two KH domains. It has been found to interact with PIWIL1 and PIWIL4 by binding to methylated arginines residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: putative RNA binding protein; RP11-98D18.8; TDRD7; tudor and KH domain containing protein; Tudor and KH domain-containing protein; tudor domain containing 2; Tudor domain-containing protein 2
Gene Aliases: 2700091C21Rik; RGD1311196; TDRD2; TDRKH
UniProt ID: (Human) Q9Y2W6, (Mouse) Q80VL1
Entrez Gene ID: (Human) 11022, (Pig) 100622748, (Mouse) 72634, (Rat) 310652
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support