Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody is stable for 24 months.
Positive Control: HeLa or MCF7 cells.Human testis or brain. Cellular Localization: Cytoplasm. Localized to cytosol and centrosome.
Specificity Comments: Participates in the primary piRNA biogenesis pathway and is required during spermatogenesis to repress transposable elements and prevent their mobilization, which is essential for the germline integrity. The piRNA metabolic process mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and govern the methylation and subsequent repression of transposons. Required for the final steps of primary piRNA biogenesis by participating in the processing of 31-37 nt intermediates into mature piRNAs. May act in pi-bodies and piP-bodies by transferring piRNA precursors or intermediates to or between these granules.
Predicted to react with Mouse and Rat
TDRKH is a protein of unknown function containing one tudor domain, and two KH domains. It has been found to interact with PIWIL1 and PIWIL4 by binding to methylated arginines residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: putative RNA binding protein; RP11-98D18.8; TDRD7; tudor and KH domain containing protein; Tudor and KH domain-containing protein; tudor domain containing 2; Tudor domain-containing protein 2
Gene Aliases: TDRD2; TDRKH
UniProt ID: (Human) Q9Y2W6
Entrez Gene ID: (Human) 11022
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support