Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: ARGPGRPAVS GFWQGALPGQ AGTSHDVKPF SQQTYAVQPP LPLPGFESPA GPAPSPSAPP APPWQGRSVA SSKLWMLEFS AFLEQQQDPD TYNKHLFVHI GQSSPSYSDP
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is preferentially expressed in the skeletal muscle, and binds to the M-CAT regulatory element found in promoters of muscle-specific genes to direct their gene expression. Alternatively spliced transcripts encoding distinct isoforms, some of which are translated through the use of a non-AUG (UUG) initiation codon, have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: related transcription enhancer factor 1B; TEA domain family member 4; TEAD-4; Transcription factor 13-like 1; Transcription factor RTEF-1; transcriptional enhancer factor 1-related; transcriptional enhancer factor 3; Transcriptional enhancer factor TEF-3
Gene Aliases: EFTR-2; hRTEF-1B; RTEF1; TCF13L1; TEAD4; TEF-3; TEF3; TEFR-1
UniProt ID: (Human) Q15561
Entrez Gene ID: (Human) 7004
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support