Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAQTVPPCEL PCKEYDVARN TGAYTSSGLA TASFRTSKYL LEEWFQNCYA RYHQAFADRD QSERQRHESQ QLATETQALA QRTQQDSTRT VGERLQDTHS WKSELQREME ALAAETNLLL AQKQRLERAL DATEVPFSIT TDNLQCRERR EHPNLVRDHV ETELLKEAEL IRNIQELLKR TIMQAVSQIR LNREHKETCE MDWSDKMEAY NIDETCGRHH SQSTEVQAHP YSTTFQESAS TPETRAKFTQ DNLCRAQRER LASANLRVLV DCILRDTSED LRLQCDAVNL AFGRRCEELE DARYKLHHHL HKTLREITDQ EHNVAALKQA IKDKEAPLHV AQTRLYLRSH RPNMELCRDA AQFRLLSEVE ELNMSLTALR EKLLEAEQSL RNLEDIHMSL EKDIAAMTNS LFIDRQKCMA HRTRYPTILQ LAGYQ
TEKT4 is structural component of ciliary and flagellar microtubules. TEKT4 forms filamentous polymers in the walls of ciliary and flagellar microtubules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tektin-4
Gene Aliases: TEKT4
UniProt ID: (Human) Q8WW24
Entrez Gene ID: (Human) 150483
Molecular Function: non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support