Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PGYPKPDMDAM EPSYELIHTQ MKTQEWDNSK SILGVQCEVQ KQLKAFVTLE RFDQLYGSTI TSCQQAPKTK KFASSGSVFG KGVKFALKDG RVTTDIISVA NEDGRR
Highest antigen sequence identity to the following orthologs - mouse 96%, rat 94%.
Involved in neural development, regulating the establishment of proper connectivity within the nervous system. Plays a role in the establishment of the anterior-posterior axis during gastrulation. Regulates the differentiation and cellular process formation of oligodendrocytes and myelination of small-diameter axons in the central nervous system (CNS) (PubMed:26188006). Promotes activation of focal adhesion kinase. May function as a cellular signal transducer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support