Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Keep as concentrated solution.
Predicted reactivity: Mouse (100%).
Positive Control: tagged-mTet1, DDDDK-mouse TET1-transfected 293T.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Dioxygenase that specifically binds methylcytosine (5mC), a minor base in mammalian DNA found in repetitive DNA elements that is crucial for retrotransposon silencing and mammalian development. Catalyzes the conversion of methylcytosine (5mC) to 5-hydroxymethylcytosine (hmC). The clear function of 5-hydroxymethylcytosine (hmC) is still unclear but it may influence chromatin structure and recruit specific factors or may constitute an intermediate component in cytosine demethylation. 5-hydroxymethylcytosine (hmC) is present in ES cells and is enriched in the brain, especially in Purkinje neurons. May play a role in the fetal development of heart, lung and brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CXXC finger 6; CXXC zinc finger 6; CXXC-type zinc finger protein 6; Leukemia-associated protein with a CXXC domain; Methylcytosine dioxygenase TET1; RP11-119F7.1; Ten-eleven translocation 1 gene protein; Ten-eleven translocation 1 gene protein homolog; ten-eleven translocation-1; tet oncogene 1; TET1 splice variant VP_DE4; TET1 splice variant VP_DE456
Gene Aliases: 2510010B09Rik; AA517754; bA119F7.1; BB001228; CXXC6; D10Ertd17e; KIAA1676; LCX; mKIAA1676; MLL-TET1; TET1; TET1-MLL
UniProt ID: (Human) Q8NFU7, (Mouse) Q3URK3
Entrez Gene ID: (Human) 80312, (Mouse) 52463
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support