Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive test controls include: Mouse liver. The target is usually found in the following locations: Cytoplasm, Nucleus.
Immunogen sequence: MDSGPVYHGD SRQLSASGVP VNGAREPAGP SLLGTGGPWR VDQKPDWEAA PGPAHTARLE DAHDLVAFSA VAEAVSSYGA LSTRLYETFN REMSREAGNN SRGPRPGPEG CSAGSEDLDT LQTALALARH
TET3, a member of the ten-eleven-translocation (TET) family of genes, is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine and is most abundantly expressed in hematopoietic cells. Unlike the related TET2 protein, mutations in TET3 have not been observed in any myeloid malignancies. TET3 has been shown to be involved in the demethylation of zygotic DNA before the first mitosis and has been suggested to be involved in the epigenetic reprogramming of the zygotic paternal DNA following natural fertilization and may also contribute to somatic cell nuclear reprogramming during animal cloning.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Methylcytosine dioxygenase TET3; MGC22014; probable methylcytosine dioxygenase TET3; putative methylcytosine dioxygenase; Tet methylcytosine deoxygenase 3 isoform; tet oncogene family member 3
Gene Aliases: B430006D22Rik; BC037432; D230004J03Rik; hCG_40738; KIAA0401; TET3
UniProt ID: (Human) Q8TBM9, (Mouse) Q8BG87
Entrez Gene ID: (Human) 200424, (Mouse) 194388
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support