Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSTTFPGLVH DAEIRHDGSN SYRLMQLGCL ESVANSTVAY SSSSPLTYST TGTEFASPYF STNHQYTPLH HQSFHYEFQH SHPAVTPDAY SLNSLHHSQQ YYQQIHHGEP TDFINLHNAR ALKSSCLDEQ RRELGCLDAY RRHDLSLMSH GSQYGMHPDQ RLLPGPSLGL AAAGADDLQG SVEAQCGLVL NGQGGVIRRG GTCVVNPTDL FCSVPGRLSL LSSTSKYKVT IAEVKRRLSP PECLNASLLG GILRRAKSKN GGRCLREKLD RLGLNLPAGR RKAANVTLLT SLVEGEALHL ARDFGYTCET EFPAKAVGEH LARQHMEQKE QTARKKMILA TKQICKEFQD LLSQDRSPLG SSRPTPILDL DIQRHLTHFS LITHGFGTPA ICAALSTFQT VLSEMLNYLE KHTTHKNGGA ADSGQGHANS EKAPLRKTSE AAVKEGKTEK TD
Sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limb and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activating enhancer binding protein 2 beta-like 1; Activating enhancer-binding protein 2-delta; AP-2 like; AP2-delta; TFAP2D; transcription factor AP-2 beta (activating enhancer binding protein 2 beta)-like 1; transcription factor AP-2 beta (activating enhancer-binding protein 2 beta)-like 1; transcription factor AP-2 delta (activating enhancer binding protein 2 delta); Transcription factor AP-2-beta-like 1; Transcription factor AP-2-delta
Gene Aliases: TFAP2BL1; TFAP2D
UniProt ID: (Human) Q7Z6R9
Entrez Gene ID: (Human) 83741
Molecular Function:
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support