Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: AEYIFSLEQE KTRLLQQNTQ LKRFIQELSG SSPKRRRAED KDEGIGSPDI WEDEKAEDLR REMIELRQQL DKERSVRMML EEQVRSLEAH MYPEKLKVIA
Transcription factors of the basic helix-loop-helix-zipper(bHLH-ZIP) family contain a basic domain, which is used for DNA binding, and HLH and ZIP domains, which are used for oligomerization. Transcription factor AP4 activates both viral and cellular genes by binding to the symmetrical DNA sequence CAGCTG.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating enhancer-binding protein 4; bHLHc41; Class C basic helix-loop-helix protein 41; Transcription factor AP-4; transcription factor AP-4 (activating enhancer-binding protein 4)
Gene Aliases: AP-4; BHLHC41; TFAP4
UniProt ID: (Human) Q01664
Entrez Gene ID: (Human) 7023
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support