Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGRRDAQLLA ALLVLGLCAL AGSEKPSPCQ CSRLSPHNRT NCGFPGITSD QCFDNGCCFD SSVTGVPWCF HPLPKQESDQ CVMEVSDRRN CGYPGISPEE CASRKCCFSN FIFEVPWCFF PKSVEDCHY
Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing and protection of the gastrointestinal epithelia. These peptides contain three intrachain disulfide bonds, forming the trefoil motif, or P-domain. SP (spasmolytic polypeptide), also designated Trefoil factor 2 (TFF2) precursor, is a trefoil protein that functions to inhibit gastrointestinal motility and gastric acid secretion. SP may also act as a structural component of the gastric mucus, possibly by stabilizing glycoproteins in the mucus gel through interactions with carbohydrate side chains. A downregulation of SP expression is associated with primary gastric cancer, and a progressive loss of this protein is likely to be involved in the early stage of the multi-step gastric carcinogenesis pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SP; Spasmolysin; Spasmolytic polypeptide; spasmolytic protein 1; Trefoil factor 2
Gene Aliases: SML1; SP; TFF2
UniProt ID: (Human) Q03403
Entrez Gene ID: (Human) 7032
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support