Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MPRSGAPKER PAEPLTPPPS YGHQPQTGSG ESSGASGDKD HLYSTVCKPR SPKPAAPAAP PFSSSSGVLG TGLCELDRLL QELNATQFNI TDEIMSQFPS SKVASGEQKE DQSEDKKRPS LPSSPSPGLP KASATSATLE LDRLMASLSD FRVQNHLPAS GPTQPPVVSS TNEGSPSPPE PTGKGSLDTM LGLLQSDLSR RGVPTQAKGL CGSCNKPIAG QVVTALGRAW HPEHFVCGGC STALGGSSFF EKDGAPFCPE CYFERFSPRC GFCNQPIRHK MVTALGTHWH PEHFCCVSCG EPFGDEGFHE REGRPYCRRD FLQLFAPRCQ GCQGPILDNY ISALSALWHP DCFVCRECFA PFSGGSFFEH EGRPLCENHF HARRGSLCAT CGLPVTGRCV SALGRRFHPD HFTCTFCLRP LTKGSFQERA GKPYCQPCFL KLFG
This gene encodes a coactivator of the androgen receptor, a transcription factor which is activated by androgen and has a key role in male sexual differentiation. The encoded protein is thought to regulate androgen receptor activity and may have a role to play in the treatment of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Androgen receptor coactivator 55 kDa protein; androgen receptor coactivator ARA55; Androgen receptor-associated protein of 55 kDa; Hic-5; Hydrogen peroxide-inducible clone 5 protein; hydrogen peroxide-inducible clone-5; Transforming growth factor beta-1-induced transcript 1 protein
Gene Aliases: ARA55; HIC-5; HIC5; TGFB1I1; TSC-5
UniProt ID: (Human) O43294
Entrez Gene ID: (Human) 7041
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support