Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LPPRERSRLL GDAKELANIL KYHIGDEILV SGGIGALVRL KSLQGDKLEV SLKNNVVSVN KEPVAEPDIM ATNGVVHVIT NVLQPPANRP QERGDELADS A
This gene encodes an RGD-containing protein that binds to type I, II and IV collagens. The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins. This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage. The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion. Mutations in this gene are associated with multiple types of corneal dystrophy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta ig-h3; beta-ig-h3; betaig-h3; Kerato-epithelin; RGD-CAP; RGD-containing collagen-associated protein; transforming growth factor beta-induced 68kDa; transforming growth factor, beta induced, 68 kDa; transforming growth factor, beta-induced, 68kD; transforming growth factor, beta-induced, 68kDa; Transforming growth factor-beta-induced protein ig-h3
Gene Aliases: 68kDa; AI181842; AI747162; Beta-ig; Big-h3; BIGH3; CDB1; CDG2; CDGG1; CSD; CSD1; CSD2; CSD3; EBMD; LCD1; TGFBI
UniProt ID: (Human) Q15582, (Mouse) P82198
Entrez Gene ID: (Human) 7045, (Mouse) 21810, (Rat) 116487
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support