Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MMDGPRSDVG RWGGNPLQPP TTPSPEPEPE PDGRSRRGGG RSFWARCCGC CSCRNAADDD WGPEPSDSRG RGSSSGTRRP GSRGSDSRRP VSRGSGVNAA GDGTIREGML VVNGVDLLSS RSDQNRREHH TDEYEYDELI VRRGQPFHML LLLSRTYESS DRITLELLIG NNPEVGKGTH VIIPVGKGGS GGWKAQVVKA SGQNLNLRVH TSPNAIIGKF QFTVRTQSDA GEFQLPFDPR NEIYILFNPW CPEDIVYVDH EDWRQEYVLN ESGRIYYGTE AQIGERTWNY GQFDHGVLDA CLYILDRRGM PYGGRGDPVN VSRVISAMVN SLDDNGVLIG NWSGDYSRGT NPSAWVGSVE ILLSYLRTGY SVPYGQCWVF AGVTTTVLRC LGLATRTVTN FNSAHDTDTS LTMDIYFDEN MKPLEHLNHD SVWNFHVWND CWMKRPDLPS GFDGWQVVDA TPQETSSGIF CCGPCSVESI KNGLVYMKYD TPFIFAEVNS DKVYWQRQDD GSFKIVYVEE KAIGTLIVTK AISSNMREDI TYLYKHPEGS DAERKAVETA AAHGSKPNVY ANRGSAEDVA MQVEAQDAVM GQDLMVSVML INHSSSRRTV KLHLYLSVTF YTGVSGTIFK ETKKEVELAP GASDRVTMPV AYKEYRPHLV DQGAMLLNVS GHVKESGQVL AKQHTFRLRT PDLSLTLLGA AVVGQECEVQ IVFKNPLPVT LTNVVFRLEG SGLQRPKILN VGDIGGNETV TLRQSFVPVR PGPRQLIASL DSPQLSQVHG VIQVDVAPAP GDGGFFSDAG GDSHLGETIP MASRGGA
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epidermal TGase; K polypeptide epidermal type I, protein-glutamine-gamma-glutamyltransferase; Protein-glutamine gamma-glutamyltransferase K; TG(K); TGase K; TGase-1; transglutaminase 1 isoform; Transglutaminase K; transglutaminase, keratinocyte; Transglutaminase-1
Gene Aliases: ARCI1; ICR2; KTG; LI; LI1; TGASE; TGK; TGM1
UniProt ID: (Human) P22735
Entrez Gene ID: (Human) 7051
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support