Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: TGN38.
IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1. and ASPP2. are activators of p53.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hTGN48; hTGN51; TGN38 homolog; TGN38B; Trans-Golgi network glycoprotein 46; Trans-Golgi network glycoprotein 48; Trans-Golgi network glycoprotein 51; Trans-Golgi network integral membrane protein 2; Trans-Golgi network integral membrane protein TGN38; trans-Golgi network protein (46, 48, 51kD isoforms); trans-golgi network protein 1; Trans-Golgi network protein 2; trans-Golgi network protein TGN51
Gene Aliases: TGN38; TGN38B; TGN41; TGN46; TGN48; TGN51; Tgoln1; TGOLN2; Ttgn1; TTGN2
UniProt ID: (Human) O43493, (Rat) P19814, (Mouse) Q62314
Entrez Gene ID: (Human) 10618, (Rat) 192152, (Mouse) 22135
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support